4-tert-Butoxyphenol 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Hydroquinone Mono-tert-butyl Ether

H26013.03 H26013.06
H26013.03EA 65.2 EUR
H26013.03 H26013.06
4-tert-Butoxyphenol 98%
4-tert-Butoxyphenol

Purity: 98% CAS No.: 2460-87-9 Molecular Formula: C10H14O2 Formula Weight: 166.22 Odour: Phenol-like Melting Point: 151-153° MDL No.: MFCD02183556

Formel: C₁₀H₁₄O₂
Molecular Weight: 166.22 g/mol
Schmelzpunkt: 151…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02183556
CAS-Nummer: 2460-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR