N-[(1R,2R)-2-Amino-1,2-diphenylethyl]-4-methylbenzolsulfonamid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (1R,2R)-(-)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine, (R,R)-TsDPEN, (1R,2R)-(-)-N-(4-Toluenesulphonyl)-1,2-diphenylethylenediamine

H26061.14 H26061.06 H26061.03
H26061.14EA 771 EUR
H26061.14 H26061.06 H26061.03
N-[(1R,2R)-2-Amino-1,2-diphenylethyl]-4-methylbenzolsulfonamid ≥98%
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulphonamide

Purity: 98+% CAS No.: 144222-34-4 Molecular Formula: C21H22N2O2S Formula Weight: 366.48 Melting Point: 127-132° Optical Rotation: -31° (c=1 in chloroform) MDL No.: MFCD02093428

Formel: C₂₁H₂₂N₂O₂S
Molecular Weight: 366.48 g/mol
Schmelzpunkt: 127…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093428
CAS-Nummer: 144222-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR