tert-Butyl hydrogen malonate 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Tert-butoxy-3-oxopropanoic acid, mono-tert-Butyl malonate

H26223.06 H26223.14
H26223.06EA 52 EUR
H26223.06 H26223.14
tert-Butyl hydrogen malonate 96%
tert-Butyl hydrogen malonate

Purity: 96% CAS No.: 40052-13-9 Molecular Formula: C7H12O4 Formula Weight: 160.17 Flash Point: 91°(195°F) Refractive Index: 1.426 Melting Point: 19-20° Boiling Point: 90°/2mm Beilstein Reference: 1765457 MDL No.: MFCD00191886

Formel: C₇H₁₂O₄
Molecular Weight: 160,17 g/mol
Siedepunkt: 90 °C (2 mmHg)
Schmelzpunkt: 19…20 °C
Dichte: 1,04
Flammpunkt: 91 °C (195 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00191886
CAS-Nummer: 40052-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR