1-Phenylethyl mercaptan 98%
Lieferant: Thermo Scientific
|
Gefahr
|
H26345.03
H26345.09
H26345.03EA
41
EUR
H26345.03
H26345.09
1-Phenylethyl mercaptan 98%
1-Phenylethyl mercaptan
Purity: 98%
CAS No.: 6263-65-6
Molecular Formula: C8H10S
Formula Weight: 138.23
Flash Point: 76°(169°F)
Refractive Index: 1.5580
UN No.: UN2810
MDL No.: MFCD00053515
Formel:
C₈H₁₀S Molecular Weight: 138,23 g/mol Dichte: 1,021 Flammpunkt: 76 °C (169 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00053515 CAS-Nummer: 6263-65-6 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...