3-[(Diethoxyphosphoryl)oxy]-1,2,3-benzotriazin-4(3H)-on 98%
Lieferant: Thermo Scientific
Achtung
Synonyme: Diethyl (4-oxobenzo[d][1,2,3]triazin-3(4H)-yl) phosphate, DEPBT, Diethyl 3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl Phosphate
H26481.06 H26481.14
H26481.06EA 57.4 EUR
H26481.06 H26481.14
3-[(Diethoxyphosphoryl)oxy]-1,2,3-benzotriazin-4(3H)-on 98%
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one
Purity: 98% CAS No.: 165534-43-0 Molecular Formula: C11H14N3O5P Formula Weight: 299.22 Storage Temperature: 2°C to 8°C Melting Point: 71-76° MDL No.: MFCD01236967
Formel: C₁₁H₁₄N₃O₅P
Molecular Weight: 299,22 g/mol
Schmelzpunkt: 71…76 °C
MDL: MFCD01236967
CAS-Nummer: 165534-43-0