(2S)-Amino(cyclohexyl)essigsäure 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Chg-OH, H-Cyclohexyl-Gly-OH, (S)-alpha-Aminocyclohexylacetic Acid
H26881.03
H26881.06
H26881.03EA
54
EUR
H26881.03
H26881.06
(2S)-Amino(cyclohexyl)essigsäure 98%
(S)-2-Amino-2-cyclohexylacetic acid
Purity: 98%
CAS No.: 14328-51-9
Molecular Formula: C8H15NO2
Formula Weight: 157.22
Melting Point:
Formel:
C₈H₁₅NO₂ Molecular Weight: 157.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311679 CAS-Nummer: 14328-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...