Methyl-1-benzyl-4-oxo-3-piperidincarboxylathydrochlorid 95%

Lieferant: Thermo Scientific

Synonyme: 1-Benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, 1-Benzyl-4-oxo-3-piperidinecarboxylic acid methyll ester hydrochloride, Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate HCl, Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride

H26930.22 H26930.14
H26930.22EA 159 EUR
H26930.22 H26930.14
Methyl-1-benzyl-4-oxo-3-piperidincarboxylathydrochlorid 95%
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride

Purity: 95% CAS No.: 3939-01-3 Molecular Formula: C14H17NO3.HCl Formula Weight: 283.75 Melting Point: ca 185° dec. MDL No.: MFCD00012799

Formel: C₁₄H₁₈ClNO₃ C₁₅H₁₉NO₃·HCl
Molecular Weight: 283,75 g/mol
Schmelzpunkt: ∼ 185 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012799
CAS-Nummer: 3939-01-3
EINECS: 223-522-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR