1-Boc-3-fluoro-4-pyrrolidinone

Lieferant: Thermo Fisher Scientific

Synonyme: tert-Butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate, 4-Fluoropyrrolidin-3-one, N-BOC protected

H27082.MD H27082.03
H27082.MDEA 174 EUR
H27082.MD H27082.03
1-Boc-3-fluoro-4-pyrrolidinone
1-Boc-3-fluoro-4-pyrrolidinone

CAS No.: 845894-03-3 Molecular Formula: C9H14FNO3 Formula Weight: 203.22 MDL No.: MFCD08274459

Formel: C₉H₁₄FNO₃
Molecular Weight: 203.213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08274459
CAS-Nummer: 845894-03-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR