D-Cysteine Hydrochloride Monohydrate 99%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: (S)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, H-D-Cys-OH·HCl·H₂O, H-D-Cys-OH.HCl
H27107.06 H27107.03
H27107.06EA 269 EUR
H27107.06 H27107.03
D-Cysteine Hydrochloride Monohydrate 99%
D-Cysteine hydrochloride monohydrate
Purity: 99% CAS No.: 207121-46-8 Molecular Formula: C3H7NO2S.HCl.H2O Formula Weight: 175.64 (157.62anhy) Storage Temperature: 2°C to 8°C Melting Point: 184-186° Optical Rotation: -5.5° (c=5 in 5N HCl) MDL No.: MFCD00150051
Formel: C₃H₇NO₂S·HCl·H₂O
Molecular Weight: 175.64 g/mol (157.62 anhy)
Schmelzpunkt: 184…186 °C
MDL: MFCD00150051
CAS-Nummer: 207121-46-8
EINECS: 251-043-9