(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol 99%
Lieferant: Thermo Scientific
Synonyme:
Boc-D-Phenylglycinol, (-)-N-Boc-D-α-phenylglycinol
H27163.03
H27163.06
H27163.03EA
86.1
EUR
H27163.03
H27163.06
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol 99%
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol
Purity: 99%
CAS No.: 102089-74-7
Molecular Formula: C13H19NO3
Formula Weight: 237.30
Melting Point: 137-141°
Optical Rotation: -40° (c=1 in chloroform)
MDL No.: MFCD00274205
Formel:
C₁₃H₁₉NO₃ Molecular Weight: 237,3 g/mol Schmelzpunkt: 137…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274205 CAS-Nummer: 102089-74-7 |