(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride 98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: H-cis-4-F-Pro-NH₂·HCl, cis-4-Fluoro-L-prolinamide hydrochloride
H27235.03 H27235.06
H27235.03EA 110 EUR
H27235.03 H27235.06
(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride 98%
(2S,4S)-4-Fluoropyrrolidine-2-carboxamide hydrochloride
Purity: 98% CAS No.: 426844-23-7 Molecular Formula: C5H9FN2O.HCl Formula Weight: 168.59 Melting Point: 231-236° dec. Optical Rotation: -45 (c=1 in water) MDL No.: MFCD07368567
Formel: C₅H₁₀ClFN₂O
Molecular Weight: 168.6 g/mol
Schmelzpunkt: 231…236 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD07368567
CAS-Nummer: 426844-23-7