(S)-2-Amino-4-phenylbutanoic acid 98%

Lieferant: Thermo Fisher Scientific

Synonyme: H-HoPhe-OH, H-Homophe-OH, (S)-2-Amino-4-phenylbutyric Acid

H27295.14 H27295.06 H27295.03
H27295.14EA 613 EUR
H27295.14 H27295.06 H27295.03
(S)-2-Amino-4-phenylbutanoic acid 98%
(S)-2-Amino-4-phenylbutanoic acid

Purity: 98% CAS No.: 943-73-7 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 300° Optical Rotation: +45° (c=1 in 3N HCl) MDL No.: MFCD00002619

Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002619
CAS-Nummer: 943-73-7
EINECS: 213-403-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR