Methyl-4-oxo-3-piperidincarboxylathydrochlorid 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 4-oxopiperidine-3-carboxylate hydrochloride, Methyl 4-Oxopiperidine-3-carboxylate HCl

H27318.14 H27318.06
H27318.14EA 301 EUR
H27318.14 H27318.06
Methyl-4-oxo-3-piperidincarboxylathydrochlorid 95%
Methyl-4-oxopiperidine-3-carboxylate hydrochloride

Purity: 95% CAS No.: 71486-53-8 Molecular Formula: C7H11NO3.HCl Formula Weight: 193.63 Melting Point: ca 175° dec. Beilstein Reference: 4273241 MDL No.: MFCD00012769

Formel: C₇H₁₂ClNO₃
Schmelzpunkt: ∼ 175 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012769
CAS-Nummer: 71486-53-8
EINECS: 275-526-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR