trans-4-Hydroxy-D-proline 97%
Lieferant: Thermo Scientific
Synonyme: (2R,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, H-D-Hyp-OH, H-trans-D-Hyp-OH
H27421.MD H27421.03
H27421.MDEA 89.6 EUR
H27421.MD H27421.03
trans-4-Hydroxy-D-proline 97%
trans-4-Hydroxy-D-proline
Purity: 97% CAS No.: 3398-22-9 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: ca 260° dec. MDL No.: MFCD00065608
Formel: C₅H₉NO₃
Molecular Weight: 131,131 g/mol
Schmelzpunkt: ∼ 260 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00065608
CAS-Nummer: 3398-22-9