4-Hexen-3-one 95% (trans-Isomere)
Lieferant: Thermo Scientific
Achtung
H27445.18 H27445.09
H27445.18EA 170 EUR
H27445.18 H27445.09
4-Hexen-3-one 95% (trans-Isomere)
4-Hexen-3-one
Purity: 98% CAS No.: 2497-21-4 Molecular Formula: C6H10O Formula Weight: 98.15 Storage Temperature: -30°C to -10°C Flash Point: 34°(93°F) Refractive Index: 1.44 Odour: Characteristic Boiling Point: 135-137° UN No.: UN1224 MDL No.: MFCD00010631
Formel: C₆H₁₀O
Molecular Weight: 98,15 g/mol
Siedepunkt: 135…137 °C
Dichte: 0,858
Flammpunkt: 34 °C (93 °F)
MDL: MFCD00010631
CAS-Nummer: 2497-21-4
EINECS: 219-681-2
ADR: 3,III