6-Brom-2H-1,4-benzoxazin-3(4H)-on 95%
Lieferant: Thermo Scientific
|
Achtung
|
H27493.09
H27493.03
H27493.09EA
252
EUR
H27493.09
H27493.03
6-Brom-2H-1,4-benzoxazin-3(4H)-on 95%
6-Bromo-2H-1,4-benzoxazin-3(4H)-one
Purity: 95%
CAS No.: 24036-52-0
Molecular Formula: C8H6BrNO2
Formula Weight: 228.04
Melting Point: 220-225°
MDL No.: MFCD00461173
Formel:
C₈H₆BrNO₂ Molecular Weight: 228,04 g/mol Schmelzpunkt: 220…225 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00461173 CAS-Nummer: 24036-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...