4-n-Pentyloxyphenylacetylene 99%

Lieferant: Thermo Scientific

Synonyme: 1-Eth-1-ynyl-4-(pentyloxy)benzene

H27555.09 H27555.03
H27555.09EA 183 EUR
H27555.09 H27555.03
4-n-Pentyloxyphenylacetylene 99%
4-n-Pentyloxyphenylacetylene

Purity: 99% CAS No.: 79887-16-4 Molecular Formula: C13H16O Formula Weight: 188.27 Flash Point: 110°(230°F) Refractive Index: 1.5310 Boiling Point: 113-115°/2mm MDL No.: MFCD00173877

Formel: C₁₃H₁₆O
Molecular Weight: 188,27 g/mol
Siedepunkt: 113…115 °C (2 mmHg)
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00173877
CAS-Nummer: 79887-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR