3-Ethyl-5-methylisoxazole-4-carboxylic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27574.09
H27574.03
H27574.09EA
128
EUR
H27574.09
H27574.03
3-Ethyl-5-methylisoxazole-4-carboxylic acid 97%
3-Ethyl-5-methylisoxazole-4-carboxylic acid
Purity: 97%
CAS No.: 17147-85-2
Molecular Formula: C7H9NO3
Formula Weight: 155.15
Melting Point: 123-125°
MDL No.: MFCD03422293
Formel:
C₇H₉NO₃ Molecular Weight: 155.15 g/mol Schmelzpunkt: 123…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03422293 CAS-Nummer: 17147-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...