2H-1,4-Benzoxazin-3(4H)-on 99%
Lieferant: Thermo Scientific
|
Achtung
|
H27622.14
H27622.06
H27622.14EA
77.1
EUR
H27622.14
H27622.06
2H-1,4-Benzoxazin-3(4H)-on 99%
2H-1,4-Benzoxazin-3(4H)-one
Purity: 99%
CAS No.: 5466-88-6
Molecular Formula: C8H7NO2
Formula Weight: 149.15
Melting Point: 173-175°
MDL No.: MFCD00158536
Formel:
C₈H₇NO₂ Molecular Weight: 149,15 g/mol Schmelzpunkt: 173…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00158536 CAS-Nummer: 5466-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...