5'-m-Terphenylboronic acid 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: [1,1':3',1''-Terphenyl]-5'-ylboronic acid

H27640.MD H27640.06 H27640.03
H27640.MDEA 24.9 EUR
H27640.MD H27640.06 H27640.03
5'-m-Terphenylboronic acid 95%
5'-m-Terphenylboronic acid

Purity: 95% CAS No.: 128388-54-5 Molecular Formula: C18H15BO2 Formula Weight: 274.13 Melting Point: 292-296° MDL No.: MFCD09953491

Formel: C₁₈H₁₅BO₂
Molecular Weight: 274,13 g/mol
Schmelzpunkt: 292…296 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09953491
CAS-Nummer: 128388-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR