3-Iodo-1-(phenylsulphonyl)indole 95%

Lieferant: Thermo Scientific

Synonyme: 3-Iodo-1-(phenylsulphonyl)-1H-indole

H27696.MD H27696.03
H27696.MDEA 50.2 EUR
H27696.MD H27696.03
3-Iodo-1-(phenylsulphonyl)indole 95%
3-Iodo-1-(phenylsulphonyl)indole

Purity: 95% CAS No.: 80360-14-1 Molecular Formula: C14H10INO2S Formula Weight: 383.20 Melting Point: 125-129° MDL No.: MFCD09037474

Formel: C₁₄H₁₀INO₂S
Molecular Weight: 383,21 g/mol
Schmelzpunkt: 125…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09037474
CAS-Nummer: 80360-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR