Methyl-4-methoxyindole-2-carboxylate 99%

Lieferant: Thermo Scientific

Synonyme: Methyl 4-methoxy-1H-indole-2-carboxylate

H27743.03 H27743.MD
H27743.03EA 102 EUR
H27743.03 H27743.MD
Methyl-4-methoxyindole-2-carboxylate 99%
Methyl-4-methoxyindole-2-carboxylate

Purity: 99% CAS No.: 111258-23-2 Molecular Formula: C11H11NO3 Formula Weight: 205.21 Melting Point: 140-142° MDL No.: MFCD00134300

Formel: C₁₁H₁₁NO₃
Molecular Weight: 205,21 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134300
CAS-Nummer: 111258-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR