Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

Lieferant: Thermo Scientific
Achtung

Synonyme: [(ppy)2IrCl]2, Dichlorotetrakis[2-(2-pyridinyl)phenyl]diiridium(III), Dichlorotetrakis(2-(2-pyridinyl)phenyl)diiridium(III)

H27752.ME H27752.MC
H27752.MEEA 420 EUR
H27752.ME H27752.MC
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

CAS No.: 92220-65-0 Molecular Formula: C44H32Cl2Ir2N4 Formula Weight: 1072.09 MDL No.: MFCD07784515

Formel: C₄₄H₃₂Cl₂Ir₂N₄
Molecular Weight: 1072,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784515
CAS-Nummer: 92220-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR