Methyl hydrogen malonate 96%
Lieferant: Thermo Scientific
|
Achtung
|
H27772.06
H27772.22
H27772.14
H27772.06EA
54.7
EUR
H27772.06
H27772.22
H27772.14
Methyl hydrogen malonate 96%
Methyl hydrogen malonate
Purity: 96%
CAS No.: 16695-14-0
Molecular Formula: HO2CCH2CO2CH3
Formula Weight: 118.09
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4295
MDL No.: MFCD00667796
Formel:
HO₂CCH₂CO₂CH₃ Molecular Weight: 118,09 g/mol Dichte: 1,128 |
MDL:
MFCD00667796 CAS-Nummer: 16695-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...