6-Nitro-1,3-benzoxazol-2(3H)-one 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Nitrobenzoxazole-2(3H)-one
H27796.06
H27796.03
H27796.06EA
76.7
EUR
H27796.06
H27796.03
6-Nitro-1,3-benzoxazol-2(3H)-one 98%
6-Nitro-1,3-benzoxazol-2(3H)-one
Purity: 98%
CAS No.: 4694-91-1
Molecular Formula: C7H4N2O4
Formula Weight: 180.12
Melting Point: 244-249°
MDL No.: MFCD00463755
Formel:
C₇H₄N₂O₄ Molecular Weight: 180,12 g/mol Schmelzpunkt: 244…249 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463755 CAS-Nummer: 4694-91-1 EINECS: 225-157-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...