2-Methyl-2-propanyl-3-oxo-1-piperazincarboxylat 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: tert-butyl 3-oxopiperazine-1-carboxylate, tert-butyl 3-oxo-1-piperazinecarboxylate, 1-Boc-3-Oxopiperazine, 4-N-BOC-2-oxo-piperazine, 4-Boc-2-Oxopiperazine

H27823.06 H27823.03
H27823.06EA 208 EUR
H27823.06 H27823.03
2-Methyl-2-propanyl-3-oxo-1-piperazincarboxylat 98%
1-Boc-3-oxopiperazine

Purity: 98% CAS No.: 76003-29-7 Molecular Formula: C9H16N2O3 Formula Weight: 200.23 Melting Point: 156-160° MDL No.: MFCD02181069

Formel: C₉H₁₆N₂O₃
Molecular Weight: 200,238 g/mol
Schmelzpunkt: 156…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02181069
CAS-Nummer: 76003-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR