7,8-Dihydroxyflavone hydrate 97%
Lieferant: Thermo Scientific
Synonyme:
7,8-Dihydroxy-2-phenyl-4H-chromen-4-one, 7,8-Dihydroxyflavone
H27852.03
H27852.06
H27852.03EA
88.9
EUR
H27852.03
H27852.06
7,8-Dihydroxyflavone hydrate 97%
7,8-Dihydroxyflavone hydrate
Purity: 97%
CAS No.: 38183-03-8
Molecular Formula: C15H10O4.xH2O
Formula Weight: 254.24(anhy)
Melting Point: 250-252°
MDL No.: MFCD00006836
Formel:
C₁₅H₁₀O₄ Molecular Weight: 254,24 g/mol Schmelzpunkt: 250…252 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006836 CAS-Nummer: 38183-03-8 EINECS: 253-812-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...