(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol ≥98%
Lieferant: Thermo Scientific
Synonyme:
(3R,4S,5R,6R)-3,4,5-tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol, 2,3,4,6-Tetra-O-benzyl-D-glucopyranose
H27888.03
H27888.06
H27888.03EA
51.1
EUR
H27888.03
H27888.06
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol ≥98%
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol
Purity: 98+%
CAS No.: 4132-28-9
Molecular Formula: C34H36O6
Formula Weight: 540.65
Storage Temperature: -30°C to -10°C
Melting Point: 150-155°
MDL No.: MFCD00066004
Formel:
C₃₄H₃₆O₆ Molecular Weight: 540,65 g/mol Schmelzpunkt: 150…155 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00066004 CAS-Nummer: 4132-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...