6-Chloro-3-pyridineacetic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(6-Chloropyridin-3-yl)acetic acid

H27966.18 H27966.09
H27966.18EA 315 EUR
H27966.18 H27966.09
6-Chloro-3-pyridineacetic acid 97%
6-Chloro-3-pyridineacetic acid

Purity: 97% CAS No.: 39891-13-9 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 172-176° MDL No.: MFCD01863172

Formel: C₇H₆ClNO₂
Molecular Weight: 171,58 g/mol
Schmelzpunkt: 172…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863172
CAS-Nummer: 39891-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR