2-Cyclohexylethanamine 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (2-Aminoethyl)cyclohexane, 2-Cyclohexylethylamine

H27987.06 H27987.03
H27987.06EA 392 EUR
H27987.06 H27987.03
2-Cyclohexylethanamine 97%
2-Cyclohexylethanamine

Purity: 97% CAS No.: 4442-85-7 Molecular Formula: C8H17N Formula Weight: 127.23 UN No.: UN2735 MDL No.: MFCD00058668

Formel: C₈H₁₇N
Molecular Weight: 127.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01734731
CAS-Nummer: 4442-85-7
EINECS: 224-673-7
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR