1,4-Bis(methoxydimethylsilyl)benzene 97%
Lieferant: Thermo Scientific
|
Achtung
|
H28165.06
H28165.03
H28165.06EA
125
EUR
H28165.06
H28165.03
1,4-Bis(methoxydimethylsilyl)benzene 97%
1,4-Bis(methoxydimethylsilyl)benzene
Purity: 97%
CAS No.: 131182-68-8
Molecular Formula: C12H22O2Si2
Formula Weight: 254.47
MDL No.: MFCD09266162
Formel:
C₁₂H₂₂O₂Si₂ Molecular Weight: 254,47 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09266162 CAS-Nummer: 131182-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...