L-Phenylalaninbenzylester -hydrochlorid 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Phe-OBzl.HCl, H-Phe-OBzl·HCl
H28407.14
H28407.06
H28407.14EA
230
EUR
H28407.14
H28407.06
L-Phenylalaninbenzylester -hydrochlorid 98%
L-Phenylalaninbenzylester -hydrochlorid
Purity: 98%
CAS No.: 2462-32-0
Molecular Formula: C16H17NO2.HCl
Formula Weight: 291.77
Storage Temperature: 2°C to 8°C
Melting Point: 197-200°
MDL No.: MFCD00043249
Formel:
C₁₆H₁₇NO₂ Molecular Weight: 291.78 g/mol Schmelzpunkt: 197…200 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00043249 CAS-Nummer: 2462-32-0 EINECS: 219-558-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...