4-Cyano-4-phenylpiperidine hydrochloride 97%

Lieferant: Thermo Scientific
Gefahr

H28414.09 H28414.03
H28414.09EA 77 EUR
H28414.09 H28414.03
4-Cyano-4-phenylpiperidine hydrochloride 97%
4-Cyano-4-phenylpiperidine hydrochloride

Purity: 97% CAS No.: 51304-58-6 Molecular Formula: C12H14N2.HCl Formula Weight: 222.71 Melting Point: 210-214° UN No.: UN3439 MDL No.: MFCD00012775

Formel: C₁₂H₁₅ClN₂
Schmelzpunkt: 210…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012775
CAS-Nummer: 51304-58-6
EINECS: 257-124-5
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR