3-Benzylbenzeneboronic acid pinacol ester 97%

Lieferant: Thermo Scientific

H28682.06 H28682.03
H28682.06EA 240 EUR
H28682.06 H28682.03
3-Benzylbenzeneboronic acid pinacol ester 97%
3-Benzylbenzeneboronic acid pinacol ester

Purity: 97% CAS No.: 1073355-05-1 Molecular Formula: C19H23BO2 Formula Weight: 294.20 Melting Point: 68-69° MDL No.: MFCD09266179

Formel: C₁₉H₂₃BO₂
Molecular Weight: 294,20 g/mol
Schmelzpunkt: 68…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09266179
CAS-Nummer: 1073355-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR