2,4,6-Trivinylboroxin pyridine complex 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Trivinylcyclotriboroxan - Pyridine Complex
H28703.06
H28703.03
H28703.06EA
451
EUR
H28703.06
H28703.03
2,4,6-Trivinylboroxin pyridine complex 95%
2,4,6-Trivinylboroxin pyridine complex
Purity: 95%
CAS No.: 95010-17-6
Molecular Formula: C6H9B3O3
Formula Weight: 161.57
Storage Temperature: 2°C to 8°C
Melting Point: 44-52°
MDL No.: MFCD03839940
Formel:
C₁₁H₁₄B₃NO₃ Molecular Weight: 240,67 g/mol Schmelzpunkt: 44…52 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03839440 CAS-Nummer: 95010-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...