N-Methyl-1-naphthylmethylamine hydrochloride 98%
Lieferant: Thermo Scientific
Achtung
Synonyme: Terbinafine Related Compound A, N-Methyl-1-(naphthalen-1-yl)methanamine hydrochloride, 1-(Methylaminomethyl)naphthalene Hydrochloride
H28719.03 H28719.09
H28719.03EA 45.1 EUR
H28719.03 H28719.09
N-Methyl-1-naphthylmethylamine hydrochloride 98%
N-Methyl-1-naphthylmethylamine hydrochloride
Purity: 98% CAS No.: 65473-13-4 Molecular Formula: C12H13N.HCl Formula Weight: 207.70 Melting Point: 190-193° MDL No.: MFCD00012555
Formel: C₁₂H₁₃N
Molecular Weight: 207,70 g/mol
Schmelzpunkt: 190…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012555
CAS-Nummer: 65473-13-4