(4-Boc-aminophenyl)boronic acid 97%
Lieferant: Thermo Scientific
Achtung
Synonyme: (4-Boc-Aminophenyl)Boronic Acid, 4-Boc-aminophenylboronic acid, (4-Boc-Aminophenyl)boronic acid, 4-(N-BOC-amino)phenylboronic acid, [4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic Acid
H28799.06 H28799.03
H28799.06EA 120 EUR
H28799.06 H28799.03
(4-Boc-aminophenyl)boronic acid 97%
(4-Boc-aminophenyl)boronic acid
Purity: 97% CAS No.: 380430-49-9 Molecular Formula: C11H16BNO4 Formula Weight: 237.06 Melting Point: ca 200° dec. MDL No.: MFCD02093054
Formel: C₁₁H₁₆BNO₄
Molecular Weight: 237,062 g/mol
Schmelzpunkt: ∼ 200 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD02093054
CAS-Nummer: 380430-49-9