(S)-Piperazine-2-carboxylic acid dihydrochloride ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2S)-piperazine-2-carboxylic acid dihydrochloride

H28972.06 H28972.03
H28972.06EA 144 EUR
H28972.06 H28972.03
(S)-Piperazine-2-carboxylic acid dihydrochloride ≥98%
(S)-Piperazine-2-carboxylic acid dihydrochloride

Purity: 98+% CAS No.: 158663-69-5 Molecular Formula: C5H10N2O2.2HCl Formula Weight: 203.07 Melting Point: ca 280° dec. Optical Rotation: -5° (c=1 in water) MDL No.: MFCD01632104

Formel: C₅H₁₂Cl₂N₂O₂
Molecular Weight: 203,07 g/mol
Schmelzpunkt: ∼ 280 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01632104
CAS-Nummer: 158663-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR