4-Benzyloxy-1-brom-2-fluorbenzol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(Benzyloxy)-1-bromo-2-fluorobenzene
H30053.06
H30053.14
H30053.06EA
61.8
EUR
H30053.06
H30053.14
4-Benzyloxy-1-brom-2-fluorbenzol 98%
4-Benzyloxy-1-brom-2-fluorbenzol
Purity: 98%
CAS No.: 185346-79-6
Molecular Formula: C13H10BrFO
Formula Weight: 281.13
Melting Point: 33-35°
MDL No.: MFCD09801041
Formel:
C₁₃H₁₀BrFO Molecular Weight: 281,13 g/mol Schmelzpunkt: 33…35 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09801041 CAS-Nummer: 185346-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...