1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 97%

Lieferant: Thermo Fisher Scientific
Achtung

H30237.MD H30237.03
H30237.MDEA 68.3 EUR
H30237.MD H30237.03
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 97%
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile

Purity: 97% CAS No.: 517918-95-5 Molecular Formula: C8H5N3 Formula Weight: 143.15 Melting Point: 226-230° MDL No.: MFCD06659684

Formel: C₈H₅N₃
Molecular Weight: 143.15 g/mol
Schmelzpunkt: 226…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659684
CAS-Nummer: 517918-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR