Carbon black ≥99% (Metall-Basis), Super P® Conductive
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Furnace black, Lamp Black
H30253.14 H30253.22
H30253.14EA 177 EUR
H30253.14 H30253.22
Carbon black ≥99% (Metall-Basis), Super P® Conductive
Carbon black
Purity: 99+% CAS No.: 1333-86-4 Molecular Formula: C Formula Weight: 12.01 Odour: Odourless UN No.: UN1325 MDL No.: MFCD00133992
Formel: C
Molecular Weight: 12.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00133992
CAS-Nummer: 1333-86-4
EINECS: 215-609-9
UN: 1325
ADR: 4.1 III