1,4-Bis(phenylethynyl)benzene 97%
Lieferant: Thermo Scientific
H30395.03
H30395.06
H30395.14
H30395.03EA
40.2
EUR
H30395.03
H30395.06
H30395.14
1,4-Bis(phenylethynyl)benzene 97%
1,4-Bis(phenylethynyl)benzene
Purity: 97%
CAS No.: 1849-27-0
Molecular Formula: C22H14
Formula Weight: 278.36
Melting Point: 176-178°
MDL No.: MFCD00160829
Formel:
C₂₂H₁₄ Molecular Weight: 278,36 g/mol Schmelzpunkt: 176…178 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00160829 CAS-Nummer: 1849-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...