2-Amino-5-chlorobenzyl alcohol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(2-Amino-5-chlorophenyl)methanol
H30531.06
H30531.14
H30531.06EA
35.4
EUR
H30531.06
H30531.14
2-Amino-5-chlorobenzyl alcohol 98%
2-Amino-5-chlorobenzyl alcohol
Purity: 98%
CAS No.: 37585-25-4
Molecular Formula: C7H8ClNO
Formula Weight: 157.60
Melting Point: 107-109°
MDL No.: MFCD00075163
Formel:
C₇H₈ClNO Molecular Weight: 157,60 g/mol Schmelzpunkt: 107…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075163 CAS-Nummer: 37585-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...