4-Fluoro-trans-β-styrylboronic acid pinacol ester 97%

Lieferant: Thermo Scientific

Synonyme: (E)-2-(4-Fluorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H30578.03 H30578.06
H30578.03EA 68.3 EUR
H30578.03 H30578.06
4-Fluoro-trans-β-styrylboronic acid pinacol ester 97%
4-Fluoro-trans-beta-styrylboronic acid pinacol ester

Purity: 97% CAS No.: 504433-86-7 Molecular Formula: C14H18BFO2 Formula Weight: 248.11 MDL No.: MFCD12546189

Formel: C₁₄H₁₈BFO₂
Molecular Weight: 248,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12546189
CAS-Nummer: 504433-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR