D-Prolinamid  98%

Lieferant: Thermo Fisher Scientific

Synonyme: H-D-Pro-NH₂, (R)-2-Pyrrolidinecarboxamide, H-D-Pro-NH2

H30901.03 H30901.06
H30901.03EA 58.3 EUR
H30901.03 H30901.06
D-Prolinamid  98%
D-Prolinamide

Purity: 98% CAS No.: 62937-45-5 Molecular Formula: C5H10N2O Formula Weight: 114.15 Melting Point: 100-103° Optical Rotation: +107° (c=2 in ethanol) MDL No.: MFCD00153457

Formel: C₅H₁₀N₂O
Molecular Weight: 114,15 g/mol
Schmelzpunkt: 100…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00153457
CAS-Nummer: 62937-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR