Boc-N-methyl-D-alanine 98%

Lieferant: Thermo Scientific

Synonyme: Boc-N-Me-Ala-OH, N-Boc-N-methyl-L-alanine

H31317.03 H31317.06
H31317.03EA 61.7 EUR
H31317.03 H31317.06
Boc-N-methyl-D-alanine 98%
Boc-N-methyl-D-alanine

Purity: 98% CAS No.: 16948-16-6 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Odour: Odourless Melting Point: 88-92° Optical Rotation: -31° (c=1 in ethanol) MDL No.: MFCD00037242

Formel: C₉H₁₇NO₄
Molecular Weight: 203,2 g/mol
Schmelzpunkt: 88…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037242
CAS-Nummer: 16948-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR