2-Amino-5-bromothiazole hydrobromide 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-5-bromothiazole monohydrobromide, 2-Amino-5-bromothiazole HBr

H31514.06 H31514.14
H31514.06EA 44.5 EUR
H31514.06 H31514.14
2-Amino-5-bromothiazole hydrobromide 97%
2-Amino-5-bromothiazole hydrobromide

Purity: 97% CAS No.: 61296-22-8 Molecular Formula: C3H3BrN2S.HBr Formula Weight: 259.95 Melting Point: ca 165° dec. MDL No.: MFCD00012712

Formel: C₃H₃BrN₂S
Molecular Weight: 259.95 g/mol
Schmelzpunkt: ∼ 165 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012712
CAS-Nummer: 61296-22-8
EINECS: 262-699-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR