Pinacyanol iodide 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Iodide, Quinaldine Blue, 1,1'-Diethyl-2,2'-carbocyanine Iodide

H31540.MD H31540.03
H31540.MDEA 46.6 EUR
H31540.MD H31540.03
Pinacyanol iodide 96%
Pinacyanol iodide

Purity: 96% CAS No.: 605-91-4 Molecular Formula: C25H25IN2 Formula Weight: 480.38 Storage Temperature: 2°C to 8°C Melting Point: 250° UN No.: UN2811 MDL No.: MFCD00011975

Formel: C₂₅H₂₅IN₂
Molecular Weight: 480,39 g/mol
Schmelzpunkt: >250 °C
MDL: MFCD00011975
CAS-Nummer: 605-91-4
EINECS: 210-099-4
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR