Pyrrolidin-2-ylacetic acid hydrochloride 97%

Lieferant: Thermo Scientific

Synonyme: Pyrrolidin-2-yl-acetic acid hydrochloride

H31749.MD H31749.03
H31749.MDEA 67.3 EUR
H31749.MD H31749.03
Pyrrolidin-2-ylacetic acid hydrochloride 97%
Pyrrolidin-2-ylacetic acid hydrochloride

Purity: 97% CAS No.: 71985-79-0 Molecular Formula: C6H11NO2.HCl Formula Weight: 165.62 Melting Point: 171-174° MDL No.: MFCD11506301

Formel: C₆H₁₂ClNO₂
Molecular Weight: 165,62 g/mol
Schmelzpunkt: 171…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11506301
CAS-Nummer: 71985-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR