Methyl-2-bromo-3-fluoropropionate 97%

Lieferant: Thermo Fisher Scientific
Gefahr

H31790.MD H31790.03
H31790.MDEA 121 EUR
H31790.MD H31790.03
Methyl-2-bromo-3-fluoropropionate 97%
Methyl-2-bromo-3-fluoropropionate

Applications:
In organic synthesis. Employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis.

Formel: C₄H₆BrFO₂
Siedepunkt: 50…54 °C (8 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD09800641
CAS-Nummer: 1537-52-6
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR